Learn More
Medicom™ SafeMask™ Artisan Procedure Earloop Mask
Catalog No. 17018281 Shop All Medicom Products
Change view
Click to view available options
Color:
Blue
ASTM Level:
ASTM Level 3
Catalog No. Color ASTM Level
17-018-281 Blue ASTM Level 3
1 options
This item is not returnable. View return policy
Catalog No. 17-018-281 Supplier Medicom™ Supplier No. 221415
Add to Cart
Add to Cart
This item is not returnable. View return policy
SafeMask™ Artisan procedure earloop mask combines a PTFE membrane with moisture-wicking technology for advanced filtration, breathability, and comfort. Ultra-lightweight, it offers a premium mask-wearing experience.
  • SafeMask™ Artisan is a multi-layered, shingle-pleat, earloop face mask with adjustable nose-piece and PTFE inner filtration layer.
  • Combines a PTFE membrane with moisture-wicking technology for advanced filtration, breathability, and comfort.
  • Ultra-lightweight, it offers a premium mask-wearing experience.
  • Procedure masks are intended to be worn by personnel during procedures to protect both the patient and personnel from transfer of microorganisms, body fluids and particulate material transfer.
  • Latex-Free.
  • ASTM F2100 standard of conformance.
  • Procedure masks are intended to be worn by personnel during procedures to protect both the patient and personnel from transfer of microorganisms, body fluids and particulate material transfer.
Specifications
Product Type Medical/Procedural Face Mask
ASTM Level ASTM Level 3
Color Blue
Fastening Type Earloops
Material Polypropylene
Size One Size
Sterility Non-sterile
Style Pleated
Certifications/Compliance ASTM F2100
Disposable Single-use
Latex Free Yes