Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medicom™ SafeMask™ Cone Face Masks

Catalog No. 19910659 Shop All Medicom Products
Change view
Click to view available options
Color:
Blue
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-910-659 Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-910-659 Supplier Medicom™ Supplier No. 2020
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Utility mask made of latex-free, porous material which helps protect the wearer from body fluids and contamination caused by exhaled micro-organisms.

  • Molded contour design increases comfort and allows for  breathability
  • Adjustable, aluminum nosepiece and stretchable headband for a custom, close fit
  • Materials and construction are soft and non-irritating
  • Comfortable to wear and alternative to shingle pleat masks

Specifications

Product Type General Purpose Face Mask
Color Blue
Fastening Type Headband
Material Polyester
Size Standard
Sterility Non-sterile
Style Cone
Disposable Single-use
For Use With (Application) Dental, industrial, medical, laboratory and safety
Latex Free Yes
Model 2020
Packaging Quantity 600 Masks/Cs.
Packaging Type Shrink wrapped
Series SafeMask™
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.