Learn More
Medicom™ SafeMask™ FreeFlow™ with Air Space™ Technology Face Masks
Catalog No. 19910712 Shop All Medicom Products
Change view
Click to view available options
Color:
Blue
Pink
Purple
White
ASTM Level:
ASTM Level 1
ASTM Level 2
ASTM Level 3
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color ASTM Level
19-910-712 Blue ASTM Level 1
19-910-711 Purple ASTM Level 1
19-910-713 Pink ASTM Level 1
19-910-714 Blue ASTM Level 2
19-910-715 Pink ASTM Level 2
19-910-718 Blue ASTM Level 3
19-910-716 Purple ASTM Level 3
19-910-719 Pink ASTM Level 3
19-910-717 White ASTM Level 3
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Catalog No. 19-910-712 Supplier Medicom™ Supplier No. 200315
Add to Cart
Add to Cart
This item is not returnable. View return policy
ASTM Level 1, 2 and 3 face masks feature innovative contoured pleats to create a unique pocket that allows air to flow freely. Ideal for those who feel claustrophobic in ordinary masks.
  • European contoured pleat design with Air Space™ technology creates unique air pocket to allow air to flow freely
  • Ideal for those who feel claustrophobic in regular masks
  • Choose from ASTM Level 1, 2 or 3
  • Unique fog-free strip integrated into the adjustable nose piece to provide clear vision when wearing protective eyewear
  • Soft, strong, sonically sealed earloops have ample length, so there is no pulling or friction
  • Comfortable fit
Specifications
Product Type Medical/Procedural Face Mask
ASTM Level ASTM Level 1
Color Blue
Fastening Type Earloops
Material Polypropylene
Size Standard
Sterility Non-sterile
Style Pleated
Certifications/Compliance ASTM F2299
Construction 3-ply
Disposable Single-use
Filter Efficiency ≥95% (PFE), ≥95% (BFE)
Latex Free Yes
Series SafeMask™ FreeFlow™
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov