Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medicom™ SafeMask™ Master Series™ Earloop Face Masks

Catalog No. 19910670 Shop All Medicom Products
Change view
Click to view available options
Color:
Blue
Pink
Purple
ASTM Level:
ASTM Level 1
ASTM Level 2
ASTM Level 3
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color ASTM Level
19-910-670 Purple ASTM Level 3
19-910-662 Pink ASTM Level 1
19-910-665 Purple ASTM Level 1
19-910-667 Blue ASTM Level 2
19-910-666 Pink ASTM Level 2
19-910-668 Blue ASTM Level 3
19-910-669 Pink ASTM Level 3
19-910-661 Blue ASTM Level 1
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Catalog No. 19-910-670 Supplier Medicom™ Supplier No. 2059
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Engineered with Simply Soft™ technology for softness and a comfortable fit without compromising protection or breathability. Available in all 3 levels of ASTM protection and in a variety of vibrant colors.

  • Variety of unique and vibrant colors
  • Simply Soft Technology™ for a smooth, luxurious and comfortable fit
  • Breathability and softness
  • Level 1, L2 and L3 protection
  • Ultrasonically sealed earloops
  • Shingle pleat design prevents fluid pooling and cross-contamination
  • High-quality media ensures optimal filtration
  • Intended to be worn during medical procedures to protect the patient and the operating personnel
  • Latex free

Specifications

Product Type Medical/Procedural Face Mask
ASTM Level ASTM Level 3
Color Purple
Fastening Type Earloops
Material Spunbond Polypropylene
Size Standard
Sterility Non-sterile
Style Pleated
Certifications/Compliance ASTM F2299, ASTM F1862
Certifications/Compliance (Europe) EN 14683
Construction 3-ply
Disposable Single-use
Filter Efficiency ≥95% (PFE), ≥95% (BFE)
For Use With (Application) Medical procedures
Latex Free Yes
Packaging Quantity 50 Masks/Box, 10 Boxes/Cs.
Pore Size 0.1 μm
Series SafeMask™ Master Series
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.