Learn More
Medicom™ SafeMask™ Duckbill Respirator
Catalog No. 19910647 Shop All Medicom Products
Change view
Click to view available options
Color: White
Catalog No.
19-910-647 Color: White
1 options
This item is not returnable. View return policy
Catalog No. 19-910-647 Supplier Medicom™ Supplier No. M2321/DEL
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
This item is not returnable. View return policy
Spacious air chamber, lightweight construction and soft inner lining ensure comfort and breathability. Adjustable nosepiece for customized fit.
  • Complies with OSHA Bloodborne Pathogens Standard when properly fitted to face and worn with proper protective eyewear
  • Maximized air chamber, lightweight construction and soft inner lining contribute to user comfort and breathability
  • Adjustable nosepiece for custom fit
  • Secure seal against most face sizes and shapes
Specifications
Color White
Product Type General Purpose Face Mask
Model M2321
Quantity 20/Pk., 8 Pk./Cs.
Sterility Non-sterile
Length (Metric) 13.1 cm