Learn More
Medicom™ SafeMask™ Premier Elite Face Masks
Catalog No. p-9327005 Shop All Medicom Products
Change view
Click to view available options
Color:
Blue
Lavender
Pink
Teal
White
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-910-720 Blue
12-888-32 Lavender
19-910-721 Pink
12-888-33 Teal
19-910-722 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-910-720 Supplier Medicom™ Supplier No. 2042
Add to Cart
Add to Cart
This item is not returnable. View return policy
ASTM Level 3 high-barrier face masks with earloops are ideal for for higher-risk patients and procedures.
  • High-barrier mask (ASTM Level 3) for examining or treating high-risk patients and for procedures involving high levels of spray, splatter, moisture and airborne particles
  • Smooth, stretchy, latex-free, ultrasonically sealed earloops prevent irritation and allow easy removal with a lower risk of self-contamination
  • Adjustable nosepiece provides a comfortable, personalized fit
  • Shingle pleat design prevents fluid pooling and cross-contamination
  • Non-woven layers will not lint, tear or shred
Specifications
Product Type Medical/Procedural Face Mask
ASTM Level ASTM Level 3
Color Blue
Fastening Type Earloops
Material Polypropylene
Size Standard
Sterility Non-sterile
Style Pleated
Certifications/Compliance ASTM F2299, ASTM F1862
Construction 3-ply
Disposable Single-use
Filter Efficiency ≥98% (PFE), ≥98% (BFE)
Latex Free Yes
Model 2042
Series SafeMask™ Premier Elite™