Description
- Integrated wraparound shield protects the eyes; ideal for treating high-risk patients and for procedures producing heavy amounts of airborne particles, moisture, spray and spatter
- Non-woven spunbond outer layer provides maximum protection against fluid penetration.
- Soft, strong, ultrasonically sealed earloops attached to outside of mask prevent pulling and irritation and allow easy removal with less risk of self-contamination.
- Shingle pleat design prevents fluid pooling and cross-contamination.
- High-quality media ensures optimal filtration and breathability.
- Adjustable, enclosed nosepiece forms a strong seal to provide customized protection and comfort.
Specifications
Specifications
| Product Type | Medical/Procedural Face Mask |
| ASTM Level | ASTM Level 3 |
| Color | Teal |
| Fastening Type | Earloops |
| Sterility | Non-sterile |
| Style | Pleated with Eyeshield |
| Disposable | Single-use |
| Latex Free | Yes |