Learn More
Medicom™ SafeMask™ Premier Elite™ ProShield Earloop Mask with Visor
Catalog No. 19910849 Shop All Medicom Products
Change view
Click to view available options
Color:
Blue
Pink
Teal
Fastening Type:
Earloops
Snaps
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Fastening Type
19-910-849 Pink Snaps
19-910-847 Teal Earloops
19-910-848 Blue Snaps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-910-849 Supplier Medicom™ Supplier No. 204216
Add to Cart
Add to Cart
This item is not returnable. View return policy
ASTM Level 3 mask with integrated anti-fog, distortion-free visor, latex-free, adjustable nosepiece and soft, strong earloops. Premium materials will not lint, tear or shred.
  • Integrated wraparound shield protects the eyes; ideal for treating high-risk patients and for procedures producing heavy amounts of airborne particles, moisture, spray and spatter
  • Non-woven spunbond outer layer provides maximum protection against fluid penetration.
  • Soft, strong, ultrasonically sealed earloops attached to outside of mask prevent pulling and irritation and allow easy removal with less risk of self-contamination.
  • Shingle pleat design prevents fluid pooling and cross-contamination.
  • High-quality media ensures optimal filtration and breathability.
  • Adjustable, enclosed nosepiece forms a strong seal to provide customized protection and comfort.
Specifications
Product Type Medical/Procedural Face Mask
ASTM Level ASTM Level 3
Color Pink
Fastening Type Snaps
Sterility Non-sterile
Style Pleated with Eyeshield
Disposable Single-use
Latex Free Yes