Learn More
Medicom™ SafeMask™ Premier Pediatric Procedure Earloop Mask, ASTM Level 1
Catalog No. 17000571 Shop All Medicom Products
Change view
Click to view available options
Pediatric L1 Masks; Packaging Quantity: 500 Masks/Cs.
Catalog No.
17-000-571 Pediatric L1 Masks; Packaging Quantity: 500 Masks/Cs.
1 options
This item is not returnable. View return policy
Catalog No. 17-000-571 Supplier Medicom™ Supplier No. 2035
  • $112.69
    $108.65 / Case of 500
    Save $4.04
    4% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
This item is not returnable. View return policy
Pediatric face masks provide ASTM Level 1 protection and enhanced comfort for children.
  • Adult-level protection in a mask made just for kids.
  • Recommended for children from 4 to 12 years of age
  • Offers a protective barrier to provide respiratory protection in procedures or situations where there is a risk of microorganism, body fluid, and particulate aerosol transfer
  • Single use, non-sterile disposable device that is recommended for use in healthcare settings with appropriate adult supervision
Specifications
Product Type General Purpose Face Mask
Color Blue
Fastening Type Earloops
Sterility Non-sterile
Style Pleated
Disposable Single-use
Latex Free Yes
Packaging Quantity 500 Masks/Cs.