Learn More
Medicom™ SafeMask™ SofSkin™ Face Masks
Catalog No. p-7182803 Shop All Medicom Products
Change view
Click to view available options
Color:
Blue
Lavender
Pink
White
ASTM Level:
ASTM Level 1
ASTM Level 3
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color ASTM Level
19-910-726 Blue ASTM Level 1
19-910-728 Lavender ASTM Level 1
19-910-727 Pink ASTM Level 1
19-910-725 White ASTM Level 1
19-910-708 Blue ASTM Level 3
19-910-710 Lavender ASTM Level 3
19-910-709 Pink ASTM Level 3
19-910-707 White ASTM Level 3
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Catalog No. 19-910-726 Supplier Medicom™ Supplier No. 2081
Add to Cart
Add to Cart
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
missing translation for 'nonReturnableMessage' missing translation for 'returnPolicyLabel'
SofSkin™ procedural face masks with earloops are ultra-soft, lightweight, non-irritating and remain cool on skin.
  • Unique cellulose-blend inner layer is ultra-soft, white, dye-free
  • Choose from ASTM Level 1 or 3
  • Adjustable nose piece
  • Unique shingle design
  • Soft, stretchy, latex-free, sonically sealed earloops
  • Comfortable and non-irritating
  • Customized fit and breathability
  • Keeps skin clean and dry
  • Will not lint, tear or shred
  • Easy removal to help prevent self-contamination
Specifications
Product Type Medical/Procedural Face Mask
ASTM Level ASTM Level 1
Color Blue
Fastening Type Earloops
Material Polypropylene
Size Standard
Sterility Non-sterile
Style Pleated
Construction 3-ply
Disposable Single-use
Filter Efficiency ≥95% (PFE), ≥95% (BFE)
Latex Free Yes
Model 2081
Quantity 50 Masks/Box, 10 Boxes/Cs., 500 Masks/Cs.
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov