Learn More
Medicom™ SafeMask Tailormade Earloop Mask
Catalog No. 19910861 Shop All Medicom Products
Change view
Click to view available options
ASTM Level:
ASTM Level 1
ASTM Level 3
Color:
Blue
Pink
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. ASTM Level Color
19-910-861 ASTM Level 1 Pink
19-910-862 ASTM Level 3 Blue
19-910-860 ASTM Level 1 Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-910-861 Supplier Medicom™ Supplier No. 2071
Add to Cart
Add to Cart
This item is not returnable. View return policy
SafeMask Tailormade Earloop Face Masks are available in 50/box, 10 boxes/Cs. and 500/Cs.
  • Dual-fit nose and chin pieces adjust to form a tight seal around the face to reduce gapping and ensure a personalized fit.
  • Soft non-woven material enhances breathability.
  • Smooth stretchy latex-free ultrasonically sealed earloops prevent irritation and allow easy removal with a lower risk of self-contamination.
Specifications
Product Type Medical/Procedural Face Mask
ASTM Level ASTM Level 1
Color Pink
Fastening Type Earloops
Sterility Non-sterile
Style Pleated
Disposable Single-use
Latex Free Yes
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov