Learn More
Medicom™ SafeMask™ Vision Procedure Earloop Mask
Catalog No. 17467292 Shop All Medicom Products
Change view
Click to view available options
SafeMask™ Vision
Catalog No.
17-467-292 SafeMask™ Vision
1 options
This item is not returnable. View return policy
Catalog No. 17-467-292 Supplier Medicom™ Supplier No. 221514
Add to Cart
Add to Cart
This item is not returnable. View return policy
SafeMask™ Vision procedure earloop mask designed with innovative filtration material and a transparent window that aids in speech reading, ensuring essential protection and patient care.
  • SafeMask™ Vision transparent window procedure earloop mask,
  • Designed with innovative filtration material, flat Omega pleat, adjustable nosepiece and a transparent window that aids in speech reading, ensuring essential protection and patient care.
  • Procedure masks are intended to be worn by personnel during procedures to protect both the patient and personnel from transfer of microorganisms, body fluids and particulate material transfer.
  • Latex-Free.
  • ASTM F2100 standard of conformance.
Specifications
Product Type Medical/Procedural Face Mask
ASTM Level ASTM Level 3
Color White
Fastening Type Earloops
Material Polypropylene
Size One Size
Sterility Non-sterile
Style Pleated
Certifications/Compliance ASTM F2100
Disposable Single-use
Latex Free Yes