Learn More
Vileda Professional™ SafePlus™ System Replacement Parts
Catalog No. 185642595 Shop All Vileda Professional Products
Change view
Click to view available options
Product Type:
Feeder for Dispenser
Flexible Lid for Dispenser
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
18-564-2595 Feeder for Dispenser
18-564-2596 Flexible Lid for Dispenser
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 18-564-2595 Supplier Vileda Professional™ Supplier No. 166931
Add to Cart
Add to Cart
This item is not returnable. View return policy
Trust the plus! The SafePlus™ System is the safe way to dispense single use microfiber wipes
  • Flexible lid is antibacterial, and self-closing to ensure the dispenser is never left open
  • Antibacterial properties of the lid are certified by a 3rd party, Hohenstein Labs
  • Lid remains antibacterial even after 80x autoclave cycles (Certified by 3rd party, Hohensteins Labs)
  • Sealed polybags wipes are packaged to fit perfectly into dispensers providing extra safety in preparation, use, and storage
Specifications
Product Type Feeder for Dispenser
Material Polypropylene
Description Replacement Part
For Use With (Equipment) SafePlus™ Systems
Sterility Non-sterile