Learn More
Andwin Scientific SafeSeal™ Microcentrifuge Tubes, Multi Color, 1.7mL, Pack of 500
SDP
Catalog No. NC9883503
Encompass_Preferred
The MµlTI™ SafeSeal™ Microcentrifuge Tubes are disposable tubes made for biotechnology applications. This product is made from precision molded polypropylene. Caps are attached onto the tubes and are specially designed to allow users to easily open the tubes, but at the same time ensure secure sample storage during boiling procedures. This product has a volume capacity of 1.7 mL and is sold in a pack of 500 multicolor tubes.
Catalog No. NC9883503 Supplier Andwin Scientific Supplier No. 53550778
Add to Cart
Add to Cart
The MµlTI™ SafeSeal™ Microcentrifuge Tubes are disposable tubes made for biotechnology applications. This product is made from precision molded polypropylene. Caps are attached onto the tubes and are specially designed to allow users to easily open the tubes, but at the same time ensure secure sample storage during boiling procedures. This product has a volume capacity of 1.7 mL and is sold in a pack of 500 multicolor tubes.