Learn More
LAMDA BIOTECH INC SafeStain 2x PCR Master Mix
Catalog No. NC3726374
Encompass_Preferred
The SafeStain™ 2X PCR Master Mix is an advanced research reagent combining PCR master mix with DNA visualization capability in a single solution. The formulation includes Taq Plus™ DNA polymerase, optimized concentrations of dNTPs, Mg2+, and PCR reaction buffer components.
Catalog No. NC3726374 Supplier LAMDA BIOTECH INC Supplier No. D139MIX
The SafeStain™ 2X PCR Master Mix is an advanced research reagent combining PCR master mix with DNA visualization capability in a single solution. The formulation includes Taq Plus™ DNA polymerase, optimized concentrations of dNTPs, Mg2+, and PCR reaction buffer components.