Learn More
LAMDA BIOTECH INC SafeStain Super-PCR Kit
Catalog No. NC3726385
Encompass_Preferred
A comprehensive PCR system combining a fluorescent dye-integrated master mix with pre-stained DNA ladder. Features non-carcinogenic green fluorescent dye for direct DNA visualization during electrophoresis.
Catalog No. NC3726385 Supplier LAMDA BIOTECH INC Supplier No. D139
A comprehensive PCR system combining a fluorescent dye-integrated master mix with pre-stained DNA ladder. Features non-carcinogenic green fluorescent dye for direct DNA visualization during electrophoresis.