Learn More
Air Science Usa Swab Drying Cabinet, Swab Dryer, 115V 60Hz
SDP
Catalog No. NC1727803
Encompass_Preferred
Safe Swab Swab Drying Cabinet, Benchtop with HEPA main filter formulated to contain any particulates or biological material that might be present on swabs, Safeswab is used to safely dry evidence swabs while protecting them from cross-contamination. Includes two 15 capacity swab drying racks. (fdc-001)
Catalog No. NC1727803 Supplier Air Science Usa Supplier No. FDC001A
Safe Swab Swab Drying Cabinet, Benchtop with HEPA main filter formulated to contain any particulates or biological material that might be present on swabs, Safeswab is used to safely dry evidence swabs while protecting them from cross-contamination. Includes two 15 capacity swab drying racks. (fdc-001)