Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Safetec™ EZ Cleans™ Kit
SDP

Catalog No. 19315617
Encompass
Encompass_Preferred
Change view
Click to view available options
Type:
Ez-Cleans Plus™ Spill Cleanup Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
19-315-617 Ez-Cleans Plus™ Spill Cleanup Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-315-617 Supplier Safetec™ Supplier No. 17121
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

  • One pair vinyl gloves
  • Odor mask
  • Red Z™ solidifier (10g pouch)
  • Scoop/scraper
  • SaniZide Plus™ germicidal wipe (8 x 8 in.)
  • Red bio-hazard waste bag (24 x 24 in.)
  • White disposal bag (30 x 36 in.)
  • Two twist ties
  • Two p.a.w.s.™ antimicrobial hand wipes (5 x 8)
  • Instructions

Kit to minimize cross-contamination

Compact, single-use kits designed for the ultimate personal protection against potentially infectious blood and body fluids during the cleanup of accidental spills. Helps reduce exposure to infectious waste as well as possible cross-contamination.

Specifications

Type Ez-Cleans Plus™ Spill Cleanup Kit
For Use With (Application) Protects against blood and body fluid spill
Includes One pair of vinyl gloves, 10 g Red Z™ solidifier, scoop or scraper, SaniZide Plus™ germicidal wipe, P.A.W.S.™ antimicrobial hand wipe, paper towel, red biohazard bag, twist tie, instructions
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.