Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Safetec™ Zorb Sheets
SDP

Catalog No. 19033120
Encompass_Preferred
Change view
Click to view available options
Dimensions (L x W):
3 x 6 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
19-033120 3 x 6 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-033120 Supplier Safetec™ Supplier No. 44004
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Absorbs and encapsulates spills containing 3% or more water

  • Tissue-based paper impregnated with Green-Z™ Biohazard Fluid Control Powder
  • Can be used in the bottom of red bags, burn boxes, specimen shipping and sharps containers
  • Help keep your employees and the public safe, as well as in compliance with the many regulations for packaging and transportation of diagnostic specimens, biological materials and other dangerous goods

Order Info

Sheets measure 3 x 6 in. (7.6 x 15.2cm). Sorbing capacity: 10cc fluid per square inch.

Specifications

Material Tissue-based Paper, Super-absorbent Polymers
Type Zorb sheet
Format 2500/Cs.
For Use With (Application) Used for employees and the public safe, as well as in compliance with the many regulations for packaging and transportation of diagnostic specimens, biological materials and other dangerous goods like bottom of red bags, burn boxes, specimen shipping
Packaging Quantity 2500/Cs.
Dimensions (L x W) 3 x 6 in.
Width (English) 3 in.
Width (Metric) 7.6 cm
Height (English) 6 in.
Height (Metric) 15.2 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.