Learn More
StatLab SafeTech Organic Vapor Monitor
SDP
Catalog No. 22021473
Change view
Click to view available options
Product Type:
Vapor Monitor
Catalog No. Product Type
22-021-473 Vapor Monitor
1 options
Catalog No. 22-021-473 Supplier StatLab Supplier No. VMBORGANICS/5
Add to Cart
Add to Cart
Vapor monitoring is important to ensure the safety of employees and comply with OSHA guidelines.
  • StatLab makes it easy to monitor vapor levels of organics in your laboratory
  • Simply send the badges in and access your results via our website
  • SafeTech Monitoring Badges for ethanol, isopropyl, and xylene exposure
  • Includes prepaid analysis for 8 hr. or 15 min. exposure readings
  • Complies with OSHA standards for vapor exposure in the workplace
  • 5/Pk.
Specifications
Product Type Vapor Monitor