Learn More
StatLab SafeTech Organics Vapor Monitor Badge: Ethanol, Isopropyl, and Xylene
SDP
Catalog No. 23763002
Change view
Click to view available options
Model:
1EA
PK6
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model
23-763-002 1EA
23-763-003 PK6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 23-763-002 Supplier StatLab Supplier No. VMBORGANICS
Add to Cart
Add to Cart
Vapor monitoring is important to ensure the safety of employees and comply with OSHA guidelines
  • SafeTech Organcis Vapor Monitor Badge will measure for ethanol, isopropyl, and xylene, it's your one stop vapor monitoring solution!
  • Simply send the badges in and access your results via our website
  • Includes prepaid analysis for 8-hour or 15-minute exposure readings
  • Complies with OSHA standards for vapor exposure in the workplace
Specifications
Detectable Analytes Organic Vapor
Product Type Organics Vapor Monitor Badge
Model 1EA