Learn More
StatLab SafeTech Organics Vapor Monitor Badge: Ethanol, Isopropyl, and Xylene
Vapor monitoring is important to ensure the safety of employees and comply with OSHA guidelines
$118.10
Specifications
| Detectable Analytes | Organic Vapor |
|---|---|
| Product Type | Organics Vapor Monitor Badge |
| Catalog Number | Mfr. No. | Model | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Model | Price | Quantity & Availability | |||||
23-763-002
![]() |
StatLab
VMBORGANICS |
1EA |
Each for $118.10
|
|
|||||
23-763-003
![]() |
StatLab
VMBORGANICS/6 |
PK6 | N/A | N/A | N/A | ||||
Description
- SafeTech Organcis Vapor Monitor Badge will measure for ethanol, isopropyl, and xylene, it's your one stop vapor monitoring solution!
- Simply send the badges in and access your results via our website
- Includes prepaid analysis for 8-hour or 15-minute exposure readings
- Complies with OSHA standards for vapor exposure in the workplace
Specifications
| Organic Vapor | |
| Organics Vapor Monitor Badge |