Learn More
MSC Chicago Hardware Anchor Shackle, Bolt Pin, 5/16 in, 1500 lb, Galvanized
Catalog No. 5000515868
Encompass
This anchor shackle is designed for various lifting and rigging applications. It features a bolt pin and a galvanized finish, offering durability and corrosion resistance. Constructed from carbon steel, it provides a reliable working load limit of 1500 lbs and meets ASME B30.26 standards.
  • Bolt pin design
  • 5/16 inch size
  • 1500 lb working load limit
  • Galvanized finish
  • Carbon steel construction
  • Meets ASME B30.26 standards
  • Heat treated shackles and pins
  • Made in USA
Catalog No. 50-005-15868 Supplier MSC Supplier No. 84002468
This anchor shackle is designed for various lifting and rigging applications. It features a bolt pin and a galvanized finish, offering durability and corrosion resistance. Constructed from carbon steel, it provides a reliable working load limit of 1500 lbs and meets ASME B30.26 standards.
  • Bolt pin design
  • 5/16 inch size
  • 1500 lb working load limit
  • Galvanized finish
  • Carbon steel construction
  • Meets ASME B30.26 standards
  • Heat treated shackles and pins
  • Made in USA