Learn More
Caresfield Safety Blood Collection Needle with Integrated Safety Shield 48/BX - 10 BX/CS
SDP
Catalog No. 22306387
Change view
Click to view available options
Color:
Black
Blue
Green
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
22-306-387 Blue
22-306-385 Green
22-306-386 Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 22-306-387 Supplier Caresfield Supplier No. 61627CS
Add to Cart
Add to Cart
The Blood Collection Needle with Integrated Safety Shield are sterile and come in different colors.
  • Hinged safety shield mechanism
  • Large, ergonomic finger grip
  • Double sheaths
  • 480 per case
Specifications
Product Type Safety Blood Collection Needle
Material Stainless Steel
Sterility Sterile
Color Blue
Disposable Single-use
Latex Free Yes