Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VacuFlo™ Safety Blood Collection Needle

Catalog No. p-9166004
Change view
Click to view available options
Gauge:
21 G
22 G
Color:
Black
Green
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Gauge Color
22-757-222 21 G Green
22-757-223 22 G Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 22-757-222 Supplier VacuFlo™ Supplier No. 607
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Uses the same safety-shield activation technique as leading brands without a change in technique. One-handed activation without a hard surface needed.

  • The same safety-shield activation technique as leading brands
  • One-handed activation without a hard surface needed

Specifications

Product Type Safety Blood Collection Needle
Material PVC
Sterility Sterile
Actuation One-handed (Manual)
Cannula Material Stainless Steel
Certifications/Compliance 510k
Color Green
Connectors Multi-Sample Luer Adapter
Disposable Single-use
For Use With (Application) Venous Multi-sample Blood Collection
For Use With (Equipment) Blood Collection Tube Holder and Blood Collection Tube
Gauge 21 G
Includes Safety Needle with Multi-sample Luer Adapter
Latex Free Yes
Length (English) 1.25 in.
Length (Metric) 3.17 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.