Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Safety Bottle Carrier

Catalog No. 16985677 Shop All Fisherbrand Products
Change view
Click to view available options
Color:
Black
Blue
Green
Red
Size:
1.5 L
4.5 L
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Size
16-985-677 Red 1.5 L
16-985-674 Green 1.5 L
16-985-676 Black 1.5 L
16-985-675 Blue 1.5 L
16-985-679 Blue 4.5 L
16-985-681 Red 4.5 L
16-985-680 Black 4.5 L
16-985-678 Green 4.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 16-985-677
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

The Safety Bottle Carrier provides a safe and reliable way to transport a variety of bottled reagents in your lab.

  • Robust Design
  • Ergonomic handle prevents hand fatigue and related accidents when carrying heavier loads
  • Handle can be pressed down to either side for easier loading and unloading
  • Centering ring stabilizes bottle in the center of carrier for safer, steadier transport
  • Carriers come in four colors with designated labeling areas for easy identification
  • Made of chemical resistant thermoplastic elastomer - PP blend

Specifications

Description Safety Bottle Carrier, 1.5 L, Red
Autoclavable Not Autoclavable
Color Red
Holds Bottles
Material Polypropylene
Positions 1
Shape Cylindrical
Stackable No
Sterility Non-sterile
For Use With (Application) Transportation of bottled regeants
For Use With (Equipment) 1.5 L bottles
Size 1.5 L
Temperature (Metric) Range -20 to 60°C
Length (Metric) 11.4 cm
Diameter (Metric) Opening 11.4 cm
Weight (Metric) .25 kg
Includes 1 Safety Bottle Carrier vessel and attachable handle
Product Type Bottle Holder
Diameter (Metric) Inner 11.4 cm
Shelves None
Height (Metric) 16.5 cm (25.4 cm Height, Handle Extended)
Array 1 x 1
Width (Metric) 11.4 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.