Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heathrow Scientific Safety Bottle Carrier

Catalog No. 14127626
Change view
Click to view available options
Color:
Black
Blue
Green
Red
Size:
1.5 L
4.5 L
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Size
14-127-626 Green 1.5 L
14-127-624 Black 1.5 L
14-127-625 Blue 1.5 L
14-127-618 Red 1.5 L
14-127-598 Black 4.5 L
14-127-604 Green 4.5 L
14-127-596 Blue 4.5 L
14-127-622 Red 4.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 14-127-626 Supplier Heathrow Scientific Supplier No. 120918
Add to Cart
Add to Cart

The Safety Bottle Carrier provides a safe and reliable way to transport a variety of bottled reagents in your lab. This carrier is ideal for any clinical and research lab setting.

  • Robust Design
  • Ergonomic handle prevents hand fatigue and related accidents when carrying heavier loads
  • Handle can be pressed down to either side for easier loading and unloading
  • Centering ring stabilizes bottle in the center of carrier for safer, steadier transport
  • Carriers come in four colors with designated labeling areas for easy identification
  • Made of chemical resistant thermoplastic elastomer - PP blend

Specifications

Autoclavable Autoclavable
Color Green
Holds Bottles
Material Polypropylene
Positions 1
Shape Cylindrical
Stackable No
Sterility Non-sterile
Size 1.5 L
Width (English) 6.5 in.
Height (English) 6.5 in.
Diameter (English) Inner 6.5 in.
Diameter (Metric) Opening 11.4 cm
Weight (Metric) 0.25 kg
Includes 1 Safety Bottle Carrier vessel and attachable handle
Product Type Safety Bottle Carrier
Shelves None
Height (Metric) 16.5 cm
Array 1 x 1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.