Learn More
Chemglass Life Sciences Chemglass Safety Bottle Carrier, 4.5 L, Red (for 4.5 L laboratory bottles)
SDP
Catalog No. NC3588482
Encompass
The Chemglass Safety Bottle Carrier is a flexible, chemical-resistant carrier designed to transport and protect 4.5 L laboratory bottles. It includes an ergonomic swing-out handle for comfortable carrying, a centering ring for bottle stability, and a recessed area for marking. The carrier is dishwasher safe and autoclavable to 121°C. Bottles are sold separately.
  • Holds 4.5 L laboratory bottles.
  • Flexible chemical-resistant plastic.
  • Autoclavable up to 121°C.
  • Dishwasher safe.
  • Ergonomic swing-out handle to reduce hand fatigue.
  • Centering ring in the base for bottle stability.
  • Recessed marking area for labels.
  • Bottles sold separately.
Catalog No. NC3588482 Supplier Chemglass Life Sciences Supplier No. CLS17105451
The Chemglass Safety Bottle Carrier is a flexible, chemical-resistant carrier designed to transport and protect 4.5 L laboratory bottles. It includes an ergonomic swing-out handle for comfortable carrying, a centering ring for bottle stability, and a recessed area for marking. The carrier is dishwasher safe and autoclavable to 121°C. Bottles are sold separately.
  • Holds 4.5 L laboratory bottles.
  • Flexible chemical-resistant plastic.
  • Autoclavable up to 121°C.
  • Dishwasher safe.
  • Ergonomic swing-out handle to reduce hand fatigue.
  • Centering ring in the base for bottle stability.
  • Recessed marking area for labels.
  • Bottles sold separately.