Learn More
Electron Microscopy Sciences Safety Bulb, Rubber pipette filler
SDP
Catalog No. 50930642
Encompass_Preferred
Rubber pipette filler. Natural rubber bulb allows safe, manual control of filling or discharging toxic liquids through a pipette by finger pressure (thus avoiding mouth contact). Guaranteed to be consistently flexible.
Catalog No. 50-930-642 Supplier Electron Microscopy Sciences Supplier No. 70982
Rubber pipette filler. Natural rubber bulb allows safe, manual control of filling or discharging toxic liquids through a pipette by finger pressure (thus avoiding mouth contact). Guaranteed to be consistently flexible.