Learn More
Hach Company Pipet Filler, Safety Bulb
Catalog No. 501995095 Shop All Hach Company Products
Encompass
Unique cone adapter allows universal pipet filling. Simply squeeze rubber bulb to charge for filling, place pipet into cone adapter and insert into sample. Release the bulb to fill. Remove bulb and cover pipet end with finger for dispensing. Easy and safe. Autoclavable.
Catalog No. 50-199-5095 Supplier Hach Company Supplier No. 1465100
Unique cone adapter allows universal pipet filling. Simply squeeze rubber bulb to charge for filling, place pipet into cone adapter and insert into sample. Release the bulb to fill. Remove bulb and cover pipet end with finger for dispensing. Easy and safe. Autoclavable.