Learn More
Andwin Scientific SAFETY BULBS 3/PK
SDP
Catalog No. NC9446592
Encompass_Preferred
The pipet bulb is designed to be compatible with all sizes of pipets. This product offers the needed suction to fill a standard glass and a plastic pipet without spillage. It is very easy to use and control. This product is sold in a pack of 3.
This item is not returnable. View return policy
Catalog No. NC9446592 Supplier Andwin Scientific Supplier No. 53497202
The pipet bulb is designed to be compatible with all sizes of pipets. This product offers the needed suction to fill a standard glass and a plastic pipet without spillage. It is very easy to use and control. This product is sold in a pack of 3.