Learn More
Electron Microscopy Sciences Safety Cabinet AMS, 30 x 45 cm
SDP
Catalog No. 501930258
Encompass_Preferred
Provide a safe environment for working with hazardous substances. A variable speed fan draws air though a HEPA filter, safely removing hazardous materials. Smooth acrylic and stainless surfaces can be thoroughly cleaned. The hinged hood tilts back to allow cleaning of the stainless base and the sides of the hood.
Size: 30 x 45 cm
Catalog No. 50-193-0258 Supplier Electron Microscopy Sciences Supplier No. 9966
Provide a safe environment for working with hazardous substances. A variable speed fan draws air though a HEPA filter, safely removing hazardous materials. Smooth acrylic and stainless surfaces can be thoroughly cleaned. The hinged hood tilts back to allow cleaning of the stainless base and the sides of the hood.
Size: 30 x 45 cm