Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Grainger Eagle Safety Cabinet Label, Flammable Keep Fire Away, Plastic, 8 1/2 in. x 14 1/2 in.

Catalog No. 502763240
Encompass_Preferred

This safety cabinet label provides clear identification and hazard warnings for safety cabinets used to store flammable materials. The rectangular sign features a prominent Flammable Keep Fire Away legend designed to alert personnel to potential risks in the area.

  • Legend: Flammable keep fire away
  • Dimensions: 21.6 cm x 36.8 cm (8 1/2 in. x 14 1/2 in.)
  • Material: Plastic
  • Color: Red legend on yellow background
  • Adhesive style: Permanent
  • Indoor use only
  • Single-sided printing

Catalog No. 50-276-3240 Supplier Grainger Supplier No. 15Y988
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This safety cabinet label provides clear identification and hazard warnings for safety cabinets used to store flammable materials. The rectangular sign features a prominent Flammable Keep Fire Away legend designed to alert personnel to potential risks in the area.

  • Legend: Flammable keep fire away
  • Dimensions: 21.6 cm x 36.8 cm (8 1/2 in. x 14 1/2 in.)
  • Material: Plastic
  • Color: Red legend on yellow background
  • Adhesive style: Permanent
  • Indoor use only
  • Single-sided printing

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.