Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Justrite™ Safety Can For Liquid Disposal

Catalog No. 19822427 Shop All Justrite Safety Group Products
Change view
Click to view available options
Capacity (English):
2 gal.
5 gal.
Color:
Red
White
Height (English):
14.8 in.
20 in.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English) Color Height (English)
19-822-427 5 gal. White 20 in.
19-822-428 2 gal. Red 14.8 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-822-427 Supplier Justrite™ Supplier No. 12772
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Increase workplace safety and improve cleaniliness with the proper safety and storage equipment. The Justrite™ Safety Can For Liquid Disposal provides a better solution for the containment of liquid flammable or corrosive waste.

High-density polyethylene (HDPE) disposal cans provide superior chemical resistance and leak protection. Spring-loaded sealed cap, normally closed, automatically vents between 3 and 5 psig (0.2 and 0.35 bar). Stainless steel handle and hardware with latch-catch system hold the cap in the open position for easy filling and pouring. Large-mouth opening features a built-in stainless steel flame arrester to guard against outside ignition. The broad-base design provides stability and resists accidental tip over.

Specifications

Product Type Safety Can
Description Safety Can for Liquid Disposal
Capacity (English) 5 gal.
Color White
For Use With (Application) Liquid Disposal
Material HDPE
Includes Stainless steel handle, faucet and flame arrester
Height (English) 20 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.