Learn More
Pyramex™ G304 Series Chemical Splash Goggles
Catalog No. 19151524 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Lens Coating:
Anti-fog, Scratch-resistant, UV-treated, Impact-resistant
Scratch-resistant, Impact-resistant, UV-treated
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Coating
19-151-524 Anti-fog, Scratch-resistant, UV-treated, Impact-resistant
19-151-523 Scratch-resistant, Impact-resistant, UV-treated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-151-524 Supplier Pyramex™ Supplier No. G304T
Add to Cart
Add to Cart
This item is not returnable. View return policy
Top shelf G304 Series Chemical Splash Goggles feature generously sized heavy duty body and lens.
  • Special one-way vent allows air to enter while keeping liquids out
  • Body tinted to reduce glare
  • Lenses provide 99.9% protection against harmful UV rays
Specifications
Lens Coating Anti-fog, Scratch-resistant, UV-treated, Impact-resistant
Certifications/Compliance ANSI Z87.1
Lens Tint Clear
Product Type Safety Goggles
Material Polycarbonate
Disposable Reusable
Lens Material Polycarbonate
Frame Material PVC
UV Protection U6
Frame Type Wrap-around
Autoclavable Not Autoclavable