Learn More
Pyramex™ Breakaway Headband Goggles
Catalog No. 191303506 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Lens Coating:
Anti-fog, Scratch-resistant
Catalog No. Lens Coating
19-130-3506 Anti-fog, Scratch-resistant
1 options
This item is not returnable. View return policy
Catalog No. 19-130-3506 Supplier Pyramex™ Supplier No. G404T
Add to Cart
Add to Cart
This item is not returnable. View return policy
Breakaway headband provides added protection and features quick fit retainer.
  • Chemical splash goggle features generously sized heavy duty body and lens with foam padding for comfort
  • Special one-way vent allows air to enter while keeping liquids out
  • Body tinted to reduce glare
  • Lenses provide 99.9% protection against harmful UV rays
Specifications
Lens Coating Anti-fog, Scratch-resistant
Certifications/Compliance ANSI Z87.1
Lens Tint Clear
Product Type Safety Goggles
Material Polycarbonate
UV Protection UV400