Description
- Manufactured from clear polycarbonate
- Scratch resistant
- 99% UVA/B absorption
Warranty and Services
90 days
Specifications
Specifications
| Certifications/Compliance | ANSI Z87.1 |
| Lens Tint | Clear |
| Product Type | Chemical-resistant Goggles |
| Material | Polycarbonate |
| UV Protection | 0.999 |