Learn More
Pyramex™ Clear Chemical Goggles
Catalog No. 19817309 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Lens Coating:
Anti-fog, Scratch-resistant
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Coating
19-817-309 Anti-fog, Scratch-resistant
19-817-308 -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-817-309 Supplier Pyramex™ Supplier No. G204T
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
This item is not returnable. View return policy
Pyramex™ Clear Chemical Goggles feature vent caps to restrict the influx of liquids.
  • Manufactured from clear polycarbonate
  • Scratch resistant
  • 99% UVA/B absorption
Warranty and Services
90 days
Specifications
Lens Coating Anti-fog, Scratch-resistant
Certifications/Compliance ANSI Z87.1
Lens Tint Clear
Product Type Chemical-resistant Goggles
Material Polycarbonate
UV Protection 0.999