Learn More
Fisherbrand™ Economy Impact Goggles
Catalog No. 19181501 Shop All Fisherbrand Products
Change view
Click to view available options
Impact Goggles
Catalog No.
19-181-501 Impact Goggles
1 options
This item is not returnable. View return policy
Catalog No. 19-181-501
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
Affordable goggles in impact style
  • Clear polycarbonate lens
  • Soft, flexible PVC body molds to facial contours for a comfortable fit
  • Adjustable headstrap ensures custom fit
  • Shield eyes from dust, mist and other airborne particles
  • Direct vents on goggle top and sides
Compliance
Meets ANSI/ISEA Z87.1-2020: Z87+ U6 standards
Specifications
Certifications/Compliance ANSI/ISEA Z87.1-2020: Z87+ U6
Lens Coating Uncoated
Lens Tint Clear
Product Type Impact Goggles
Ventilation Details Direct vents on both sides and top
Material Polycarbonate
Frame Material PVC