Learn More
Pyramex™ Safety Choice Perforated Goggles
Catalog No. 19817306 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Lens Coating:
Scratch-resistant, Impact-resistant, UV-treated
Catalog No. Lens Coating
19-817-306 Scratch-resistant, Impact-resistant, UV-treated
1 options
This item is not returnable. View return policy
Catalog No. 19-817-306 Supplier Pyramex™ Supplier No. G201
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
Perforated Safety Choice Perforated Goggles provide excellent ventilation. Lenses provide 99.9% protection against harmful UV rays.
  • Perforated
  • Manufactured from clear polycarbonate
  • Scratch resistant
  • 99% UVA/B absorption
Warranty and Services
90 Days
Specifications
Lens Coating Scratch-resistant, Impact-resistant, UV-treated
Certifications/Compliance ANSI Z87.1
Lens Tint Clear
Product Type Perforated Goggles
Material Polycarbonate
Disposable Reusable
Lens Material Polycarbonate
Frame Material PVC
UV Protection U6
Frame Type Wrap-around
Autoclavable Not Autoclavable