Learn More
Pyramex™ Ventless Safety Goggle
Catalog No. 191303507 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Lens Coating:
Anti-fog, Scratch-resistant
Catalog No. Lens Coating
19-130-3507 Anti-fog, Scratch-resistant
1 options
This item is not returnable. View return policy
Catalog No. 19-130-3507 Supplier Pyramex™ Supplier No. G200T
Add to Cart
Add to Cart
This item is not returnable. View return policy
Ventless goggle provides protection against odor vapors.
Lenses provide 99.9% protection against harmful UV rays
Specifications
Lens Coating Anti-fog, Scratch-resistant
Certifications/Compliance ANSI Z87.1
Lens Tint Clear
Product Type Perforated Goggles
Material Polycarbonate