Learn More
Chemglass Life Sciences Clamp, Safety, Adjustable, Fits Joint Sizes 45/35 & 45/50
SDP
Catalog No. CG145C09
Encompass_Preferred
Clamp, Safety, Adjustable, Fits Joint Sizes 45/35 & 45/50. The safety clip is a two piece adjustable clamp used to secure standard taper joints. Made of polyoxymethylene, the clip has excellent chemical resistance. They will not corrode or scratch the glass. Can be used to 150°C. Sold in packages of 10. PLEASE NOTE: THESE CLAMPS CAN BE USED WITH STANDARD TAPER JOINTS THAT HAVE A CG-136 STYLE PTFE SLEEVE BETWEEN THEM.
Catalog No. CG145C09 Supplier Chemglass Life Sciences Supplier No. CG145C09
Clamp, Safety, Adjustable, Fits Joint Sizes 45/35 & 45/50. The safety clip is a two piece adjustable clamp used to secure standard taper joints. Made of polyoxymethylene, the clip has excellent chemical resistance. They will not corrode or scratch the glass. Can be used to 150°C. Sold in packages of 10. PLEASE NOTE: THESE CLAMPS CAN BE USED WITH STANDARD TAPER JOINTS THAT HAVE A CG-136 STYLE PTFE SLEEVE BETWEEN THEM.