Learn More
Research Products International Corp Hybridization Bottle, Safety Coated Glass Bottles with Cap and PTFE Liner, 35 x 300mm
SDP
Catalog No. 50212499
Encompass
Heavy wall borosilicate glass bottles designed for use in standard hybridization ovens. Fit all rotisseries that accommodate 35mm diameter bottles. Bottles feature a leakproof polypropylene closure with a PTFE/Silicone liner. Bottles are also available with a slip resistant plastic coating.
Catalog No. 50-212-499 Supplier Research Products International Corp Supplier No. 246007
Heavy wall borosilicate glass bottles designed for use in standard hybridization ovens. Fit all rotisseries that accommodate 35mm diameter bottles. Bottles feature a leakproof polypropylene closure with a PTFE/Silicone liner. Bottles are also available with a slip resistant plastic coating.