Learn More
Omnimed Inc Safety Control Seals W Numbers 100 Pack (Yellow)
SDP
Supplier:  Omnimed Inc 484107Y
Encompass_Preferred
The Omnimed Safety Control Seals with Numbers have a brightly colored plastic body with a flexible locking piece which threads into the body. These seals are designed for a narrow slot opening. Each is imprinted with a unique tracking number and can be broken without tools. Seals can be used on cabinets where contents need to be controlled. They provide a clear visual signal and also serve as a deterrent to prevent unauthorized cabinet access. Seals are sold in 100 count packages and come in red, green or yellow.
Catalog No. 50-105-9177