Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TerraCycle™ Safety Equipment & Protective Gear Zero Waste Box™
SDPSCI_ED

Catalog No. S475652
Change view
Click to view available options
Dimensions (L x W x H):
11 x 11 x 20 in.
11 x 11 x 40 in.
15 x 15 x 37 in.
Capacity (Metric):
39.66 L
79.31 L
136.42 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W x H) Capacity (Metric)
S475652 11 x 11 x 20 in. 39.66 L
S475653 11 x 11 x 40 in. 79.31 L
S475654 15 x 15 x 37 in. 136.42 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options

These boxes are not suitable for medical waste. This includes any red bag waste (i.e. material contaminated with blood or bodily fluids that originate from health care facilities, hospitals, physicians' offices, dental offices, blood banks, veterinary clinics, research laboratories, etc.) The zero waste box system does not accept household items that may be considered hazardous waste. Do not ship dripping packages.

Not available in Hawaii, Alaska, and Puerto Rico.

Catalog No. S475652 Supplier TerraCycle™ Supplier No. CS58S
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

These boxes are not suitable for medical waste. This includes any red bag waste (i.e. material contaminated with blood or bodily fluids that originate from health care facilities, hospitals, physicians' offices, dental offices, blood banks, veterinary clinics, research laboratories, etc.) The zero waste box system does not accept household items that may be considered hazardous waste. Do not ship dripping packages.

Not available in Hawaii, Alaska, and Puerto Rico.

Recycle Safety Equipment & Protective Gear. Each box includes signage, a built-in liner, shipping to and from your location, and processing of the materials.

  • WHAT YOU CAN RECYCLE
    • Use this Zero Waste Box to recycle all brands and sizes of disposable safety equipment and protective gear.
    • This includes:
      • Ear plugs
      • Beard nets
      • Hair nets
      • Gloves
      • Safety glasses
      • Disposable Garments
      • Masks
    • Do not send in medical waste. This includes material contaminated with blood or bodily fluids that originate from health care facilities, hospitals, physicians' offices, dental offices, blood banks, veterinary clinics, research laboratories, etc.

Specifications

Capacity (Metric) 39.66 L
Color Brown, Green, White
Dimensions (L x W x H) 11 x 11 x 20 in.
Autoclavable Not Autoclavable
Material Cardboard
Product Type Recycling Box
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.