Learn More
MSC MCR Safety Safety Glasses, Anti-Fog, Polycarbonate, Clear, ANSI Z87.1+
Catalog No. 502835335
Encompass
These full-framed safety glasses are designed for optimal visibility and protection. They feature clear, anti-fog polycarbonate lenses and a durable orange polycarbonate frame with non-adjustable bayonet temples. Meeting ANSI Z87.1+ standards, these glasses offer a wraparound style with integrated side shields for comprehensive eye protection in various environments.
  • Anti-fog coatings improve visibility under hot and humid conditions.
  • Polycarbonate lenses provide impact resistance.
  • Clear lenses offer unobstructed vision.
  • Full-framed design ensures durability.
  • Wraparound style with side shields provides comprehensive eye protection.
  • Meets ANSI Z87.1+ safety standards.
  • Non-adjustable bayonet temples for a secure fit.
Catalog No. 50-283-5335 Supplier MSC Supplier No. 87524419
These full-framed safety glasses are designed for optimal visibility and protection. They feature clear, anti-fog polycarbonate lenses and a durable orange polycarbonate frame with non-adjustable bayonet temples. Meeting ANSI Z87.1+ standards, these glasses offer a wraparound style with integrated side shields for comprehensive eye protection in various environments.
  • Anti-fog coatings improve visibility under hot and humid conditions.
  • Polycarbonate lenses provide impact resistance.
  • Clear lenses offer unobstructed vision.
  • Full-framed design ensures durability.
  • Wraparound style with side shields provides comprehensive eye protection.
  • Meets ANSI Z87.1+ safety standards.
  • Non-adjustable bayonet temples for a secure fit.