Learn More
MSC Bouton Optical Safety Glasses, Clear, Anti-Scratch, FogLess 3Sixty, Rimless
Catalog No. 502835449
Encompass
These safety glasses feature a clear polycarbonate lens with advanced anti-scratch and FogLess 3Sixty™ anti-fog coatings, ensuring clear vision in demanding conditions. Designed with a lightweight, rimless frame, they provide 99.99% UV protection and are engineered for all-day user comfort and a secure fit.
  • Lightweight, rimless design provides all-day user comfort.
  • Flexible bayonet temples with rubber pads ensure a secure, non-slip fit.
  • Adjustable rubber nosepiece offers a comfortable fit.
  • FogLess 3Sixty™ enhanced hydrophobic action resists lens fogging.
  • Anti-scratch coating extends eyewear life.
  • Clear polycarbonate lens provides excellent visibility.
  • Offers 99.99% UV protection.
  • Meets ANSI Z87.1+ and CSA Z94.3 standards.
Catalog No. 50-283-5449 Supplier MSC Supplier No. 33698747
Add to Cart
Add to Cart
These safety glasses feature a clear polycarbonate lens with advanced anti-scratch and FogLess 3Sixty™ anti-fog coatings, ensuring clear vision in demanding conditions. Designed with a lightweight, rimless frame, they provide 99.99% UV protection and are engineered for all-day user comfort and a secure fit.
  • Lightweight, rimless design provides all-day user comfort.
  • Flexible bayonet temples with rubber pads ensure a secure, non-slip fit.
  • Adjustable rubber nosepiece offers a comfortable fit.
  • FogLess 3Sixty™ enhanced hydrophobic action resists lens fogging.
  • Anti-scratch coating extends eyewear life.
  • Clear polycarbonate lens provides excellent visibility.
  • Offers 99.99% UV protection.
  • Meets ANSI Z87.1+ and CSA Z94.3 standards.