Learn More
MSC Pyramex Provoq Safety Glasses, Polycarbonate, ANSI Z87.1, 1 Each
Catalog No. 5000688794
Encompass
These safety glasses are designed for superior protection and comfort in various working environments. Constructed from durable polycarbonate, they offer essential impact resistance and meet rigorous safety standards for high-impact applications.
  • Meets ANSI/ISEA Z87.1-2020 and CAN/CSA Z94.3-07 high impact standards
  • Features 99% UVA/B/C protection
  • Equipped with a soft rubber nosepiece for a comfortable fit
  • Includes rubber-tipped temples to provide a non-slip, secure fit
  • Lightweight design suitable for extended wear
  • Includes scratch-resistant lens coating
Catalog No. 50-006-88794 Supplier MSC Supplier No. 99478638
These safety glasses are designed for superior protection and comfort in various working environments. Constructed from durable polycarbonate, they offer essential impact resistance and meet rigorous safety standards for high-impact applications.
  • Meets ANSI/ISEA Z87.1-2020 and CAN/CSA Z94.3-07 high impact standards
  • Features 99% UVA/B/C protection
  • Equipped with a soft rubber nosepiece for a comfortable fit
  • Includes rubber-tipped temples to provide a non-slip, secure fit
  • Lightweight design suitable for extended wear
  • Includes scratch-resistant lens coating