Learn More
Grainger Condor Enticer Series Safety Glasses, Clear, Frameless, Scratch-Resistant, 1 ea
Catalog No. NC0017821
Encompass_Preferred
These safety glasses are designed to protect eyes from flying debris and splashes, featuring a lightweight, frameless design for comfortable, long-term wear. The glasses include padding at the nose bridge to reduce pressure and improve fit.
  • Clear polycarbonate lenses with scratch-resistant and anti-fog coatings
  • Frameless, wraparound frame style for clear peripheral vision
  • Nylon arms with bayonet-style tips for a secure fit
  • Complies with ANSI Z87.1-2015 standards
  • Provides 99.99% UVA and UVB protection
  • Operating temperature range of -5°C to +50°C (23°F to 122°F)
This item is not returnable. View return policy
Catalog No. NC0017821 Supplier Grainger Supplier No. 4VCE9
These safety glasses are designed to protect eyes from flying debris and splashes, featuring a lightweight, frameless design for comfortable, long-term wear. The glasses include padding at the nose bridge to reduce pressure and improve fit.
  • Clear polycarbonate lenses with scratch-resistant and anti-fog coatings
  • Frameless, wraparound frame style for clear peripheral vision
  • Nylon arms with bayonet-style tips for a secure fit
  • Complies with ANSI Z87.1-2015 standards
  • Provides 99.99% UVA and UVB protection
  • Operating temperature range of -5°C to +50°C (23°F to 122°F)