Learn More
MSC Safety Glasses, 3M SecureFit 3700, Orange Polycarbonate, Scratch-Resistant, 1 ea
Catalog No. 5000793611
Encompass
The 3M SecureFit 3700 Series Safety Glasses feature a frameless design with scratch-resistant, orange polycarbonate lenses. Designed for eye protection, these glasses incorporate 3M Pressure Diffusion Temple Technology for enhanced comfort and meet ANSI Z87.1-2015 and CSA Z94.3 safety standards.
  • Frameless design for a wide field of vision
  • Scratch-resistant lens coating for durability
  • Orange polycarbonate lenses provide UV protection
  • Universal size with non-adjustable temples
  • Polycarbonate frame material
  • Meets ANSI Z87.1-2015 and CSA Z94.3 standards
  • Features Pressure Diffusion Temple Technology
Catalog No. 50-007-93611 Supplier MSC Supplier No. 96198213
The 3M SecureFit 3700 Series Safety Glasses feature a frameless design with scratch-resistant, orange polycarbonate lenses. Designed for eye protection, these glasses incorporate 3M Pressure Diffusion Temple Technology for enhanced comfort and meet ANSI Z87.1-2015 and CSA Z94.3 safety standards.
  • Frameless design for a wide field of vision
  • Scratch-resistant lens coating for durability
  • Orange polycarbonate lenses provide UV protection
  • Universal size with non-adjustable temples
  • Polycarbonate frame material
  • Meets ANSI Z87.1-2015 and CSA Z94.3 standards
  • Features Pressure Diffusion Temple Technology